Learn More
2,5-Dimethoxybenzonitrile, 98%
Product Code. 11412578
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11412578
Brand: Thermo Scientific Alfa Aesar A15868.14
Request Bulk or Custom Format
CAS: 5312-97-0 | C9H9NO2 | 163.176 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5312-97-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00016375
InChI Key HWAMEJIMXIXLIH-UHFFFAOYSA-N
PubChem CID 79200
IUPAC Name 2,5-dimethoxybenzonitrile
SMILES COC1=CC(=C(C=C1)OC)C#N
Specifications
Melting Point 80°C to 84°C
Quantity 25 g
UN Number UN3439
Beilstein 2691937
Synonym benzonitrile, 2,5-dimethoxy, 2,5-dimethoxybenzenecarbonitrile, pubchem13109, acmc-1an8e, benzonitrile,2,5-dimethoxy, 2-cyano-1,4-dimethoxybenzene, 2,5-dimethoxybenzonitrile, labotest-bb lt00848082, 2-cyanohydroquinone dimethyl ether
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material 2,5-Dimethoxybenzonitrile