accessibility menu, dialog, popup

UserName

CAS RN 5312-97-0

H3COOCH3N

CAS RN 5312-97-0

IUPAC Name: 2,5-dimethoxybenzonitrile
Synonyms: 2,5-dimethoxybenzonitrile
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: HWAMEJIMXIXLIH-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results