missing translation for 'onlineSavingsMsg'
Learn More

2,5-Deoxyfructosazine-13C4, TRC

Product Code. 30360244
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360244 5 mg 5mg
30354786 1 mg 1mg
30354444 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360244 Supplier Toronto Research Chemicals Supplier No. TRCD2358125MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246815-27-9
Molecular Formula C813C4H20N2O7
Molecular Weight (g/mol) 308.27
Synonym (1R,2S,3R)-1-[5-[(2S,3R)-2,3,4-Trihydroxybutyl]-2-pyrazinyl]-1,2,3,4-butanetetrol-13C4, 2-(D-arabino-Tetrahydroxybutyl)-5-(D-erythro-2,3,4-trihydroxybutyl)pyrazine-13C4, NSC 270912-13C4
SMILES OC[C@@H](O)[C@@H](O)C[13c]1[13cH]n[13c]([13cH]n1)[C@@H](O)[C@H](O)[C@H](O)CO

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H20N2O7/c15-4-9(18)8(17)1-6-2-14-7(3-13-6)11(20)12(21)10(19)5-16/h2-3,8-12,15-21H,1,4-5H2/t8-,9+,10+,11+,12+/m0/s1/i2+1,3+1,6+1,7+1
Formula Weight 308.1405
Chemical Name or Material 2,5-Deoxyfructosazine-13C4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.