CAS RN 1246815-27-9
CAS RN 1246815-27-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,5-Deoxyfructosazine-13C4, TRC
CAS: 1246815-27-9 Molecular Formula: C813C4H20N2O7 Molecular Weight (g/mol): 308.27 Synonym: (1R,2S,3R)-1-[5-[(2S,3R)-2,3,4-Trihydroxybutyl]-2-pyrazinyl]-1,2,3,4-butanetetrol-13C4,2-(D-arabino-Tetrahydroxybutyl)-5-(D-erythro-2,3,4-trihydroxybutyl)pyrazine-13C4,NSC 270912-13C4 SMILES: OC[C@@H](O)[C@@H](O)C[13c]1[13cH]n[13c]([13cH]n1)[C@@H](O)[C@H](O)[C@H](O)CO