missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride, Mikromol™
Description
[2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | (2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy)acetic Acid Dihydrochloride |
| CAS | 83881-54-3 |
| Analyte or Component Names | (2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy)acetic Acid Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 N2 O3 . 2 Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Cetirizine Dihydrochloride Imp. F (EP), Cetirizine Imp. F (EP), [2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride, Cetirizine Dihydrochloride Imp. F (EP) as Dihydrochloride, Acetic acid, 2-[2-[4-(diphenylmethyl)-1-piperazinyl]ethoxy]-, hydrochloride (1:2), Acetic acid, [2-[4-(diphenylmethyl)-1-piperazinyl]ethoxy]-, dihydrochloride (9CI), Cetirizine Dihydrochloride Impurity F as Dihydrochloride, Cetirizine Impurity F as Dihydrochloride |
| InChI Formula | InChI=1S/C21H26N2O3.2ClH/c24-20(25)17-26-16-15-22-11-13-23(14-12-22)21(18-7-3-1-4-8-18)19-9-5-2-6-10-19;;/h1-10,21H,11-17H2,(H,24,25);2*1H |
| SMILES | O=C(COCCN1CCN(C(C2=CC=CC=C2)C3=CC=CC=C3)CC1)O.[2HCl] |
| IUPAC Name | 2-[2-(4-benzhydrylpiperazin-1-yl)ethoxy]acetic acid;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?