missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitro-6-(1-methylheptyl)phenol, TRC

Product Code. 30133322
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133322 1 mg 1mg
30134790 25 mg 25mg
30010897 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133322 Supplier Toronto Research Chemicals Supplier No. TRCD4923881MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3687-22-7
Formula Weight 296.1372
InChI Formula InChI=1S/C14H20N2O5/c1-3-4-5-6-7-10(2)12-8-11(15(18)19)9-13(14(12)17)16(20)21/h8-10,17H,3-7H2,1-2H3
IUPAC Name 2,4-dinitro-6-octan-2-ylphenol
Molecular Formula C14 H20 N2 O5
Molecular Weight (g/mol) 296.32
SMILES CCCCCCC(C)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]
Synonym 2-(1-Methylheptyl)-4,6-dinitrophenol, 2,4-DNOP, 2,4-Dinitro-6-(1-methylheptyl)phenol, 2,4-Dinitro-6-(1'-methylheptyl)phenol, 2-(2-Hydroxy-3,5-dinitrophenyl)octane, DNOP, Meptyldinocap-phenol
Chemical Name or Material Meptyldinocap-phenol

Specifications

Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.