Learn More
2,4-Difluoro-3-methylbenzylamine, 98%, Thermo Scientific™
Product Code. 15444737
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444737 5 g 5g
15454737 250 mg 250mg
15434737 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15444737 Supplier Thermo Scientific Alfa Aesar Supplier No. H32060.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 847502-90-3
Molecular Formula C8H9F2N
Molecular Weight (g/mol) 157.164
MDL Number MFCD08275293
InChI Key LPMVSOQQHWTDRH-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methylbenzylamine, 1-2,4-difluoro-3-methylphenyl methanamine, 2,4-difluoro-3-methylphenyl methanamine, benzenemethanamine,2,4-difluoro-3-methyl
PubChem CID 11469203
IUPAC Name (2,4-difluoro-3-methylphenyl)methanamine
SMILES CC1=C(C=CC(=C1F)CN)F
Specifications
Boiling Point 42°C (1 mmHg)
Refractive Index 1.495
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 157.16
Percent Purity 98%
Chemical Name or Material 2,4-Difluoro-3-methylbenzylamine