accessibility menu, dialog, popup

UserName

CAS RN 847502-90-3

CH3FH2NF

CAS RN 847502-90-3

IUPAC Name: 1-(2,4-difluoro-3-methylphenyl)methanamine
Synonyms: 1-(2,4-difluoro-3-methylphenyl)methanamine
Molecular Weight (g/mol): 157.16
Molecular Formula: C8H9F2N
InChi Key: LPMVSOQQHWTDRH-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(CN)=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results