Learn More
2,4-Diaminotoluene, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4-Diaminotoluene |
| CAS | 95-80-7 |
| Analyte or Component Names | 2,4-Diaminotoluene |
| Shipping Condition | +20°C |
| Molecular Formula | C7 H10 N2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Toluene-2,4-diamine (7CI,8CI), 4-Methyl-1,3-benzenediamine, 1,3-Diamino-4-methylbenzene, 2,4-Diamino-1-methylbenzene, 2,4-Tolylenediamine, 2-Methyl-1,5-benzenediamine, 3-Amino-4-methylaniline, 4-Methyl-1,3-diaminobenzene, 4-Methyl-1,3-phenylenediamine, 4-Methyl-m-phenylenediamine, Brown for Fur T, C.I. Oxidation Base 35, Eucanine GB, Fouramine J, Fourrine 94, Fourrine M, Nako TMT, Pelagol Grey J, Pelagol J, Pontamine Developer TN, Renal MD, TDA, TDA (diamine), Zoba GKE, m-Toluenediamine, m-Tolylenediamine |
| InChI Formula | InChI=1S/C7H10N2/c1-5-2-3-6(8)4-7(5)9/h2-4H,8-9H2,1H3 |
| SMILES | Cc1ccc(N)cc1N |
| IUPAC Name | 4-methylbenzene-1,3-diamine |
| Show More |