Learn More
2,4-Diaminodiphenylamine, 98%
Product Code. 11475596
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475596 1 g 1g
11485596 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11475596 Supplier Thermo Scientific Alfa Aesar Supplier No. B20735.03
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
CAS: 136-17-4 | C12H13N3 | 199.257 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 136-17-4
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD00025285
InChI Key VOSLIUIVGWBSOK-UHFFFAOYSA-N
Synonym 2,4-diaminodiphenylamine, diphenylamine, 2,4-diamino, n1-phenylbenzene-1,2,4-triamine, 1,2,4-benzenetriamine, n'-phenyl, unii-45dn363j8c, n sup 1-phenyl-1,2,4-benzenetriamine, 1,2,4-benzenetriamine, n sup 1-phenyl, diphenylamine,4-diamino, 1,4-benzenetriamine, n1-phenyl, 3-13-00-00552 beilstein handbook reference
PubChem CID 8683
IUPAC Name 1-N-phenylbenzene-1,2,4-triamine
SMILES C1=CC=C(C=C1)NC2=C(C=C(C=C2)N)N
Specifications
Melting Point 129°C to 131°C
Quantity 1 g
UN Number UN2811
Formula Weight 199.26
Percent Purity 98%
Chemical Name or Material 2,4-Diaminodiphenylamine