accessibility menu, dialog, popup

UserName

CAS RN 136-17-4

IUPAC Name: N1-phenylbenzene-1,2,4-triamine
Synonyms: N1-phenylbenzene-1,2,4-triamine
Molecular Weight (g/mol): 199.26
Molecular Formula: C12H13N3
InChi Key: VOSLIUIVGWBSOK-UHFFFAOYSA-N
SMILES: NC1=CC=C(NC2=CC=CC=C2)C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results