missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Di(dodecylthiomethyl)-6-methylphenol, Dr. Ehrenstorfer
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2,4-Di(dodecylthiomethyl)-6-methylphenol |
| CAS | 110675-26-8 |
| Analyte or Component Names | 2,4-Di(dodecylthiomethyl)-6-methylphenol |
| Shipping Condition | +20°C |
| Molecular Formula | C33 H60 O S2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Phenol, 2,4-bis[(dodecylthio)methyl]-6-methyl-, 2,4-Bis[(dodecylthio)methyl]-6-methylphenol, 2,4-Bis(dodecylsulfanylmethyl)-6-methylphenol, 2,4-Bis(dodecylthiomethyl)-6-methylphenol, 2,4-Bis(n-dodecylthiomethyl)-6-methylphenol, 2,4-Di(dodecylthiomethyl)-6-methylphenol, 4,6-Bis(dodecylthiomethyl)-o-cresol, Antage HP 500, Antioxidant 1726, HP 500, Irganox 1726, RC 1726 |
| InChI Formula | InChI=1S/C33H60OS2/c1-4-6-8-10-12-14-16-18-20-22-24-35-28-31-26-30(3)33(34)32(27-31)29-36-25-23-21-19-17-15-13-11-9-7-5-2/h26-27,34H,4-25,28-29H2,1-3H3 |
| SMILES | CCCCCCCCCCCCSCc1cc(C)c(O)c(CSCCCCCCCCCCCC)c1 |
| IUPAC Name | 2,4-bis(dodecylsulfanylmethyl)-6-methylphenol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction