missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Oxo-2,3-dihydrobenzofuran-5-yl)ethyl methanesulfonate, TRC

Product Code. 30425316
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
This item is not returnable. View return policy

Product Code. 30425316

Brand: Toronto Research Chemicals TRCO8553052.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(3-Oxo-2,3-dihydrobenzofuran-5-yl)ethyl methanesulfonate
Analyte or Component Names 2-(3-Oxo-2,3-dihydrobenzofuran-5-yl)ethyl methanesulfonate
Shipping Condition Room Temperature
Molecular Formula C11 H12 O5 S
InChI Formula InChI=1S/C11H12O5S/c1-17(13,14)16-5-4-8-2-3-11-9(6-8)10(12)7-15-11/h2-3,6H,4-5,7H2,1H3
Solution Type Neat
SMILES CS(=O)(=O)OCCc1ccc2OCC(=O)c2c1
IUPAC Name 2-(3-oxo-1-benzofuran-5-yl)ethyl methanesulfonate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 256.275
Quantity 2.5 mg
Formula Weight 256.041
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.