Learn More
2,3-Dibromopyridine, 97%
Product Code. 11398896
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11398896
Brand: Thermo Scientific Alfa Aesar L19527.06
Request Bulk or Custom Format
CAS: 13534-89-9 | C5H3Br2N | 236.894 g/mol
It is employed as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Moisture sensitive. Store in dry condition. Keep oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13534-89-9
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.894
MDL Number MFCD00234014
InChI Key SLMHHOVQRSSRCV-UHFFFAOYSA-N
Synonym pyridine, 2,3-dibromo, dibromopyridine, 2,3-dibromo pyridine, pubchem5402, acmc-209c0b, 2,3-dibromopyridine, ksc492s9r, tpc-py084, slmhhovqrssrcv-uhfffaoysa, abbypharma ap-12-5128
PubChem CID 817102
IUPAC Name 2,3-dibromopyridine
SMILES C1=CC(=C(N=C1)Br)Br
Specifications
Melting Point 58°C to 60°C
Boiling Point 249°C to 250°C
Quantity 5 g
Beilstein 109828
Solubility Information Insoluble in water.
Formula Weight 236.9
Percent Purity 97%
Chemical Name or Material 2,3-Dibromopyridine