accessibility menu, dialog, popup

UserName

CAS RN 13534-89-9

BrNBr

CAS RN 13534-89-9

IUPAC Name: 2,3-dibromopyridine
Synonyms: 2,3-dibromopyridine
Molecular Weight (g/mol): 236.89
Molecular Formula: C5H3Br2N
InChi Key: SLMHHOVQRSSRCV-UHFFFAOYSA-N
SMILES: BrC1=CC=CN=C1Br
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results