missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dehydro-3,4-dihydro Ivermectin (Mixture of Diastereomers)>85%, TRC

Product Code. 30005591
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005591 50 mg 50mg
30133576 250 mg 250mg
30132988 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005591 Supplier Toronto Research Chemicals Supplier No. TRCD22941050MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C48H74O14
Molecular Weight (g/mol) 875.09
Synonym (4S)-2,3-Didehydro-5-O-demethyl-3,4,22,23-tetrahydro-avermectin A1a
SMILES [H][C@@]1(O[C@H]2[C@H](C)O[C@@]([H])(O[C@H](/C(C)=C/C[C@]3([H])C[C@H]4C[C@@]5(O[C@]([H])([C@@](CC)([H])C)[C@@H](C)CC5)O3)[C@@H](C)/C=C/C=C6CO[C@@]7([H])[C@]\6(O)C(C(O4)=O)=CC([H])(C)[C@H]7O)C[C@@H]2OC)O[C@@H](C)[C@H](O)[C@@H](OC)C1

Specifications

Analyte or Component Names 2,3-Dehydro-3,4-dihydro Ivermectin (Mixture of Diastereomers)
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Chemical Name or Material 2,3-Dehydro-3,4-dihydro Ivermectin (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.