missing translation for 'onlineSavingsMsg'
Learn More

2-(3-chlorophenyl)-1-methyl-6-oxopiperidine-3-carboxylic acid, TRC

Product Code. 30359129
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359129 100 mg 100mg
30349441 10 mg -
30368726 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359129 Supplier Toronto Research Chemicals Supplier No. TRCC610718100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1394041-72-5
Molecular Formula C13H14NO3Cl
Molecular Weight (g/mol) 267.7
IUPAC Name 2-(3-chlorophenyl)-1-methyl-6-oxopiperidine-3-carboxylic acid
SMILES CN1C(C(CCC1=O)C(=O)O)c2cccc(Cl)c2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H14ClNO3/c1-15-11(16)6-5-10(13(17)18)12(15)8-3-2-4-9(14)7-8/h2-4,7,10,12H,5-6H2,1H3,(H,17,18)
Formula Weight 267.0662
Chemical Name or Material 2-(3-chlorophenyl)-1-methyl-6-oxopiperidine-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.