CAS RN 1394041-72-5
CAS RN 1394041-72-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(3-chlorophenyl)-1-methyl-6-oxopiperidine-3-carboxylic acid, TRC
CAS: 1394041-72-5 Molecular Formula: C13H14NO3Cl Molecular Weight (g/mol): 267.7 IUPAC Name: 2-(3-chlorophenyl)-1-methyl-6-oxopiperidine-3-carboxylic acid SMILES: CN1C(C(CCC1=O)C(=O)O)c2cccc(Cl)c2