Learn More
2,3,5,6-Tetrafluoro-4-hydroxypyridine, 97%, Thermo Scientific™
Product Code. 15406567
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406567 250 mg 250mg
15496557 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15406567 Supplier Thermo Scientific Alfa Aesar Supplier No. H33712.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2693-66-5
Molecular Formula C5HF4NO
Molecular Weight (g/mol) 167.06
MDL Number MFCD00981885
InChI Key OYRHEJZOLQEJFB-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoropyridin-4-ol, 4-hydroxy-2,3,5,6-tetrafluoropyridine, tetrafluoropyridin-4-ol, 2,3,5,6-tetrafluoro-4-pyridinol, 2,3,5,6-tetrafluoro-4-hydroxypyridine, tetrafluoropyridine-4-ol, 2,3,5,6-tetrafluoro-4-hydroxy-pyridine, 2,3,5,6-tetrafluoropyridin-4 1h-one, 4 1h-pyridinone, 2,3,5,6-tetrafluoro
PubChem CID 12127311
IUPAC Name 2,3,5,6-tetrafluoro-1H-pyridin-4-one
SMILES FC1=C(F)C(=O)C(F)=C(F)N1
Tekniske data
Melting Point 95°C to 97°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 167.06
Percent Purity 97%
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-hydroxypyridine
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.