accessibility menu, dialog, popup

UserName

CAS RN 2693-66-5

NHOFFFF

CAS RN 2693-66-5

IUPAC Name: 2,3,5,6-tetrafluoro-1,4-dihydropyridin-4-one
Synonyms: 2,3,5,6-tetrafluoro-1H-pyridin-4-one
Molecular Weight (g/mol): 167.06
Molecular Formula: C5HF4NO
InChi Key: OYRHEJZOLQEJFB-UHFFFAOYSA-N
SMILES: FC1=C(F)C(=O)C(F)=C(F)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results