missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethanamine Dioxalate, Mikromol™
Description
2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethanamine Dioxalate
Specifications
Specifications
| Chemical Name or Material | 2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethanamine Dioxalate |
| CAS | 78441-63-1 |
| Analyte or Component Names | 2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethanamine Dioxalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H17 N3 S2 . 2 C2 H2 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethanamine Dioxalate, Nizatidine Imp. D (EP) as Dioxalate, 2-Thiazolemethanamine, 4-[[(2-aminoethyl)thio]methyl]-N,N-dimethyl-, ethanedioate (1:2), Nizatidine Impurity D as Dioxalate |
| InChI Formula | InChI=1S/C9H17N3S2.2C2H2O4/c1-12(2)5-9-11-8(7-14-9)6-13-4-3-10;2*3-1(4)2(5)6/h7H,3-6,10H2,1-2H3;2*(H,3,4)(H,5,6) |
| SMILES | CN(C)Cc1nc(CSCCN)cs1.OC(=O)C(=O)O.OC(=O)C(=O)O |
| IUPAC Name | 2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfanyl]ethanamine;oxalic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?