missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Benzyl-1H-indol-5-yl)-2-(1H-indol-5-yl)-N-methylethane-1-sulfonamide, TRC
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | 2-(1-Benzyl-1H-indol-5-yl)-2-(1H-indol-5-yl)-N-methylethane-1-sulfonamide |
| Analyte or Component Names | 2-(1-Benzyl-1H-indol-5-yl)-2-(1H-indol-5-yl)-N-methylethane-1-sulfonamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H25 N3 O2 S |
| InChI Formula | InChI=1S/C26H25N3O2S/c1-27-32(30,31)18-24(20-7-9-25-22(15-20)11-13-28-25)21-8-10-26-23(16-21)12-14-29(26)17-19-5-3-2-4-6-19/h2-16,24,27-28H,17-18H2,1H3 |
| Solution Type | Neat |
| SMILES | CNS(=O)(=O)CC(c1ccc2[nH]ccc2c1)c3ccc4c(ccn4Cc5ccccc5)c3 |
| IUPAC Name | 2-(1-benzylindol-5-yl)-2-(1H-indol-5-yl)-N-methylethanesulfonamide |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 443.561 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction