Learn More
2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide, Mikromol™
Product Code. 30461199
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461199 4 x 25 mg 100mg
30051044 100 mg 100mg
30052166 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30461199 Supplier Mikromol Supplier No. MM0084.2000254AMP
 Restricted Item
Estimated Shipment: 02-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide
CAS 30763-03-2
Analyte or Component Names 2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide
Shipping Condition Room Temperature
Molecular Formula C9 H8 N2 O4 S
Certifications/Compliance ISO 17025
Synonym 2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide
InChI Formula InChI=1S/C9H8N2O4S/c10-8(12)5-11-9(13)6-3-1-2-4-7(6)16(11,14)15/h1-4H,5H2,(H2,10,12)
Solution Type Neat
SMILES NC(=O)CN1C(=O)c2ccccc2S1(=O)=O
IUPAC Name 2-(1,1,3-trioxo-1,2-benzothiazol-2-yl)acetamide
Molecular Weight (g/mol) 240.24
Quantity 4 x 25 mg
Formula Weight 240.0205 g/mol