Learn More
(1S)-(-)-Camphorsulfonylimine, 98+%
Product Code. 11435756
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11435756
Brand: Thermo Scientific Alfa Aesar B20823.14
Request Bulk or Custom Format
CAS: 60886-80-8 | C10H15NO2S | 213.30 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60886-80-8
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.30
MDL Number MFCD00013315,MFCD00064576
InChI Key ZAHOEBNYVSWBBW-UHFFFAOYNA-N
Synonym 1s,7s-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.0<1,5> dec-4-ene-3,3-dione, 1s,7s-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? dec-4-ene-3,3-dione
PubChem CID 45925640
IUPAC Name 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
SMILES CC1(C)C2CCC11CS(=O)(=O)N=C1C2
Specifications
Melting Point 226°C to 228°C
Quantity 25 g
Beilstein 85296
Formula Weight 213.3
Percent Purity ≥98%
Chemical Name or Material (1S)-(-)-Camphorsulfonylimine