CAS RN 60886-80-8
CAS RN 60886-80-8
IUPAC Name:
10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
Synonyms:
10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
Molecular Weight (g/mol):
213.3
Molecular Formula:
C10H15NO2S
InChi Key:
ZAHOEBNYVSWBBW-UHFFFAOYNA-N
SMILES:
CC1(C)C2CCC11CS(=O)(=O)N=C1C2
1
–
1
of
1
results
1
(1S)-(-)-Camphorsulfonylimine, 98+%
CAS: 60886-80-8 Molecular Formula: C10H15NO2S Molecular Weight (g/mol): 213.30 MDL Number: MFCD00013315,MFCD00064576 InChI Key: ZAHOEBNYVSWBBW-UHFFFAOYNA-N Synonym: 1s,7s-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.0<1,5> dec-4-ene-3,3-dione,1s,7s-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? dec-4-ene-3,3-dione PubChem CID: 45925640 IUPAC Name: 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione SMILES: CC1(C)C2CCC11CS(=O)(=O)N=C1C2