Learn More
(1S)-2,2-Dimethylcyclopropanecarboxamide, Mikromol™
Product Code. 30052250
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052250 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052250 Supplier Mikromol Supplier No. MM1092.200025
 Restricted Item
Estimated Shipment: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1S)-2,2-Dimethylcyclopropanecarboxamide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1S)-2,2-Dimethylcyclopropanecarboxamide
CAS 75885-58-4
Analyte or Component Names (1S)-2,2-Dimethylcyclopropanecarboxamide
Shipping Condition Room Temperature
Molecular Formula C6 H11 N O
Certifications/Compliance ISO 9001
Synonym Cyclopropanecarboxamide, 2,2-dimethyl-, (1S)-, Cyclopropanecarboxamide, 2,2-dimethyl-, (S)-, (1S)-2,2-Dimethylcyclopropanecarboxamide, (+)-2,2-Dimethylcyclopropanecarboxamide, (S)-2,2-Dimethylcyclopropanecarboxamide, S-(+)-2,2-Dimethylcyclopropane-1-carboxamide, S-(+)-2,2-Dimethylcyclopropanecarboxamide
InChI Formula InChI=1S/C6H11NO/c1-6(2)3-4(6)5(7)8/h4H,3H2,1-2H3,(H2,7,8)/t4-/m1/s1
SMILES CC1(C)C[C@@H]1C(=O)N
IUPAC Name (1S)-2,2-dimethylcyclopropane-1-carboxamide
Molecular Weight (g/mol) 113.1576
Quantity 25 mg
Formula Weight 113.0841 g/mol