Learn More
(1R,3S)-3-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopentanemethanol, TRC
Product Code. 30137767
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137767 50 mg 50mg
17959619 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137767 Supplier Toronto Research Chemicals Supplier No. TRCA60379550MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 208762-35-0
Molecular Formula C14 H20 N6 O
Molecular Weight (g/mol) 288.35
Synonym [(1R,3S)-3-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopentyl]methanol, Cyclopentanemethanol, 3-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, (1R,3S)-, (1R,3S)-3-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopentanemethanol, Abacavir Sulfate Imp. E (Ph. Int.), Abacavir Sulfate Imp E (EP)
IUPAC Name [(1R,3S)-3-[2-amino-6-(cyclopropylamino)purin-9-yl]cyclopentyl]methanol
SMILES Nc1nc(NC2CC2)c3ncn([C@H]4CC[C@@H](CO)C4)c3n1
Specifications
Analyte or Component Names (1R,3S)-3-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopentanemethanol
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H20N6O/c15-14-18-12(17-9-2-3-9)11-13(19-14)20(7-16-11)10-4-1-8(5-10)6-21/h7-10,21H,1-6H2,(H3,15,17,18,19)/t8-,10+/m1/s1
Solution Type Neat
Industry Type Forensics
Quantity 50 mg
Formula Weight 288.1699
Chemical Name or Material (1R,3S)-3-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopentanemethanol