missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-1-(3,4-Dihydroxyphenyl)-2-methylaminoethanesulphonic Acid (Adrenaline beta-Sulphonate), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(1R)-1-(3,4-Dihydroxyphenyl)-2-methylaminoethanesulphonic Acid (Adrenaline beta-Sulphonate)
Specifications
Specifications
| Chemical Name or Material | Adrenaline beta-Sulphonate |
| CAS | 78995-75-2 |
| Analyte or Component Names | Adrenaline beta-Sulphonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H13 N O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1R)-1-(3,4-Dihydroxyphenyl)-2-methylaminoethanesulphonic Acid, Adrenaline beta-Sulphonate, Adrenaline Impurity F |
| InChI Formula | InChI=1S/C9H13NO5S/c1-10-5-9(16(13,14)15)6-2-3-7(11)8(12)4-6/h2-4,9-12H,5H2,1H3,(H,13,14,15)/t9-/m0/s1 |
| SMILES | CNC[C@@H](c1ccc(O)c(O)c1)S(=O)(=O)O |
| IUPAC Name | (1R)-1-(3,4-dihydroxyphenyl)-2-(methylamino)ethanesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction