missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-1-(1-Naphthyl)ethylamine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(1R)-1-(1-Naphthyl)ethylamine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (1R)-1-(1-Naphthyl)ethylamine Hydrochloride |
| CAS | 82572-04-1 |
| Analyte or Component Names | (1R)-1-(1-Naphthyl)ethylamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H13 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | (1R)-1-(1-Naphthyl)ethylamine Hydrochloride |
| InChI Formula | InChI=1S/C12H13N.ClH/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11;/h2-9H,13H2,1H3;1H/t9-;/m1./s1 |
| SMILES | Cl.C[C@@H](N)c1cccc2ccccc12 |
| IUPAC Name | (1R)-1-naphthalen-1-ylethanamine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction