Learn More
IBA Lifesciences Strep-Tactin™ Sepharose™ Column
Product Code. 12886436 Shop All IBA Lifesciences Products
Change view
Click to view available options
Capacity (Metric):
0.2 mL
1 mL
Unit Size:
1mL
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12886436 1 mL 1mL
12836446 0.2 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12886436 Supplier IBA Lifesciences Supplier No. 21202001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Strep-Tactin™ Sepharose™ column is a Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™ Sepharose™ columns contains 4% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • Due to the low concentrated agarose, it is particularly suitable for large proteins, especially if the affinity to Strep-Tactin™XT 4Flow™ is too high.
  • The wide-meshed polymer allows for the simple attachment of larger proteins to the stationary phase.
  • To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended. 
Specifications
Capacity (Metric) 1 mL
For Research Use Only.