missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Droperidol 1.0 mg/ml in Dimethyl Sulfoxide, LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Droperidol 1.0 mg/ml in Dimethyl Sulfoxide
Specifications
Specifications
| Chemical Name or Material | Droperidol |
| CAS | 548-73-2 |
| Analyte or Component Names | Droperidol |
| Shipping Condition | Ice Pack (-20°C) |
| Molecular Formula | C22 H22 F N3 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Droperidol, 1-[1-[4-(4-Fluorophenyl)-4-oxobutyl]-1,2,3,6-tetrahydropyridin-4-yl]-1,3-dihydro-2 H-benzimidazol-2-one |
| InChI Formula | InChI=1 S/C22H22FN3O2/c23-17-9-7-16(8-10-17)21(27)6-3-13-25-14-11-18(12-15-25)26-20-5-2-1-4-19(20)24-22(26)28/h1-2,4-5,7-11 H,3,6,12-15H2,(H,24,28) |
| SMILES | Fc1ccc(cc1)C(=O)CCCN2CCC(=CC2)N3C(=O)Nc4ccccc34 |
| IUPAC Name | 3-[1-[4-(4-fluorophenyl)-4-oxobutyl]-3,6-dihydro-2 H-pyridin-4-yl]-1 H-benzimidazol-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction