missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmethylchlorpromazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Desmethylchlorpromazine Hydrochloride 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Desmethylchlorpromazine Hydrochloride |
| CAS | 3953-65-9 |
| Analyte or Component Names | Desmethylchlorpromazine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H17 Cl N2 S . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | 3-(2-Chloro-10 H-phenothiazin-10-yl)-N-methylpropan-1-amine Hydrochloride (Desmethylchlorpromazine Hydrochloride), Chlorpromazine Hydrochloride Imp. D (EP), Chlorpromazine Imp. D (EP), 3-(2-Chloro-10 H-phenothiazin-10-yl)-N-methylpropan-1-amine Hydrochloride, Chlorpromazine Hydrochloride Imp. D (EP) as Hydrochloride, Desmethylchlorpromazine Hydrochloride, Chlorpromazine Hydrochloride Impurity D as Hydrochloride, Chlorpromazine Impurity D as Hydrochloride |
| InChI Formula | InChI=1 S/C16H17ClN2S.ClH/c1-18-9-4-10-19-13-5-2-3-6-15(13)20-16-8-7-12(17)11-14(16)19;/h2-3,5-8,11,18 H,4,9-10H2,1H3;1 H |
| SMILES | Cl.CNCCCN1c2ccccc2Sc3ccc(Cl)cc13 |
| IUPAC Name | 3-(2-chlorophenothiazin-10-yl)-N-methylpropan-1-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction