missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorpromazine Sulphoxide 1.0 mg/ml in Methanol, LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Chlorpromazine Sulphoxide 1.0 mg/ml in Methanol
Specifications
Specifications
| Chemical Name or Material | Chlorpromazine Sulfoxide |
| CAS | 969-99-3 |
| Analyte or Component Names | Chlorpromazine Sulfoxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H19 Cl N2 O S |
| Certifications/Compliance | ISO 17034 |
| Synonym | Chlorpromazine Hydrochloride Imp. A (EP), Chlorpromazine Imp. A (EP), 3-(2-Chloro-10 H-phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide, Chlorpromazine Sulfoxide, Chlorpromazine Sulphoxide, Chlorpromazine Hydrochloride Impurity A, Chlorpromazine Impurity A |
| InChI Formula | InChI=1 S/C17H19ClN2OS/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)22(21)17-9-8-13(18)12-15(17)20/h3-4,6-9,12 H,5,10-11H2,1-2H3 |
| SMILES | CN(C)CCCN1c2ccccc2S(=O)c3ccc(Cl)cc13 |
| IUPAC Name | 3-(2-chloro-5-oxophenothiazin-10-yl)-N,N-dimethylpropan-1-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction