missing translation for 'onlineSavingsMsg'
Learn More

Bufotenine 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30463967
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30463967

Brand: LoGiCal LGCAMP1360.1011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Bufotenine 1.0 mg/ml in Acetonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bufotenine
CAS 487-93-4
Analyte or Component Names Bufotenine
Shipping Condition Room Temperature
Molecular Formula C12 H16 N2 O
Certifications/Compliance ISO 17034
Synonym 3-[2-(Dimethylamino)ethyl]-1 H-indol-5-ol, 3-[2-(Dimethylamino)ethyl]-indol-5-ol, 3-(2-Dimethylaminoethyl)indol-5-ol, 3-(beta-Dimethylaminoethyl)-5-hydroxyindole, 5-Hydroxy-N,N-dimethyltryptamine, Bufotenin, Bufotenine, Cinobufotenine, Dimethylserotonin, M
InChI Formula InChI=1 S/C12H16N2O/c1-14(2)6-5-9-8-13-12-4-3-10(15)7-11(9)12/h3-4,7-8,13,15 H,5-6H2,1-2H3
SMILES CN(C)CCc1c[nH]c2ccc(O)cc12
IUPAC Name 3-[2-(dimethylamino)ethyl]-1 H-indol-5-ol
Solvent or Matrix Acetonitrile
Industry Type Forensics
Molecular Weight (g/mol) 204.27
Quantity 1 mL
Formula Weight 204.1263 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.