missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-D cholinium 100 μg/mL in Water, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 2,4-D Cholinium |
| CAS | 1048373-72-3 |
| Analyte or Component Names | 2,4-D Cholinium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H5 Cl2 O3 . C5 H14 N O |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2,4-D choline salt, 2-Hydroxy-N,N,N-trimethylethanaminium 2-(2,4-dichlorophenoxy)acetate |
| InChI Formula | InChI=1S/C8H6Cl2O3.C5H14NO/c9-5-1-2-7(6(10)3-5)13-4-8(11)12;1-6(2,3)4-5-7/h1-3H,4H2,(H,11,12);7H,4-5H2,1-3H3/q;+1/p-1 |
| SMILES | C[N+](C)(C)CCO.[O-]C(=O)COc1ccc(Cl)cc1Cl |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction