missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibenzylpiperazine.2HCl, 1mg/ml in Acetonitrile/Water : 1/1 (as free base), Lipomed

Product Code. 30464053
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30464053

Brand: Lipomed LPMDBZ1578HC1LAH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

1,4-Dibenzylpiperazine.2HCl, 1mg/ml in Acetonitrile/Water : 1/1 (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Dibenzylpiperazine Dihydrochloride
CAS 2298-55-7
Analyte or Component Names 1,4-Dibenzylpiperazine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2 . 2 Cl H
Concentration 1 mg/mL
Synonym Piperazine, 1,4-bis(phenylmethyl)-, hydrochloride (1:2), Piperazine, 1,4-dibenzyl-, dihydrochloride (7 CI,8 CI)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.Cl.C(N1CCN(Cc2ccccc2)CC1)c3ccccc3
IUPAC Name 1,4-dibenzylpiperazine;dihydrochloride
Solvent or Matrix Acetonitrile/Water
Industry Type Pharmaceutical
Molecular Weight (g/mol) 339.3
Quantity 1 mL
Formula Weight 338.1317 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.