missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human UBE2M/Ubc12 Protein

Product Code. 18461712 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18461712 1mg 1mg
18408161 500 μg 500µg
18489771 5 μg 5µg
18425012 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18461712 Supplier Novus Biologicals™ Supplier No. NBP2351161mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P61081
For Use With (Application) PAGE
Formulation A 0.2 um filtered concentrated solution in 50 mM HEPES, pH 6.5, with 125 mM NaCl, 10 % Glycerol, 1 mM DTT.
Gene ID (Entrez) 9040
Molecular Weight (g/mol) M.W. (Theoretical): 20.9 kDa
Quantity 1mg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Endotoxin Concentration Less than 0.1 EU/μg of UBE2M/Ubc12 as determined by LAL method.
Gene Alias EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog
Gene Symbol UBE2M
Research Category Biologically Active Proteins
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein UBE2M/Ubc12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.