Learn More
17β-Hydroxy-6-methylideneandrosta-1,4-dien-3-one (17β-Hydroxyexemestane), Mikromol
Description
17β-Hydroxy-6-methylideneandrosta-1,4-dien-3-one (17β-Hydroxyexemestane), Mikromol
Specifications
Specifications
| Chemical Name or Material | 17beta-Hydroxy Exemestane |
| CAS | 122370-91-6 |
| Analyte or Component Names | 17beta-Hydroxy Exemestane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H26 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Androsta-1,4-dien-3-one, 17-hydroxy-6-methylene-, (17β)-, (17β)-17-Hydroxy-6-methyleneandrosta-1,4-dien-3-one, 17-Hydroexemestane, Hydroexemestane, 17β-Hydroxy Exemestane, 17β-Hydroxyexemestane |
| InChI Formula | InChI=1S/C20H26O2/c1-12-10-14-15-4-5-18(22)20(15,3)9-7-16(14)19(2)8-6-13(21)11-17(12)19/h6,8,11,14-16,18,22H,1,4-5,7,9-10H2,2-3H3/t14-,15-,16-,18-,19+,20-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CC(=C)C4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2O |
| Show More |