missing translation for 'onlineSavingsMsg'
Learn More

16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione, Mikromol™

Product Code. 17980946
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980946 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980946 Supplier Mikromol Supplier No. MM0204.02
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 16 alpha,17-[(1RS)-Ethylidenebis(oxy)]-11 beta,21-dihydroxypregna-1,4-diene-3,20-dione
CAS 1040085-98-0
Analyte or Component Names 16 alpha,17-[(1RS)-Ethylidenebis(oxy)]-11 beta,21-dihydroxypregna-1,4-diene-3,20-dione
Shipping Condition Room Temperature
Molecular Formula C23 H30 O6
Certifications/Compliance ISO 9001
Synonym 16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione, Budesonide Imp. B (EP), Budesonide Impurity B
InChI Formula InChI=1S/C23H30O6/c1-12-28-19-9-16-15-5-4-13-8-14(25)6-7-21(13,2)20(15)17(26)10-22(16,3)23(19,29-12)18(27)11-24/h6-8,12,15-17,19-20,24,26H,4-5,9-11H2,1-3H3/t12?,15-,16-,17-,19+,20+,21-,22-,23+/m0/s1
SMILES CC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
IUPAC Name 16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione
Molecular Weight (g/mol) 402.48
Quantity 50 mg
Formula Weight 402.2042 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.