missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11alpha-Hydroxyprogesterone, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
11alpha-Hydroxyprogesterone
Specifications
Specifications
| Chemical Name or Material | 11a-Hydroxyprogesterone |
| CAS | 80-75-1 |
| Analyte or Component Names | 11a-Hydroxyprogesterone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H30 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 11alpha-Hydroxyprogesterone |
| InChI Formula | InChI=1S/C21H30O3/c1-12(22)16-6-7-17-15-5-4-13-10-14(23)8-9-20(13,2)19(15)18(24)11-21(16,17)3/h10,15-19,24H,4-9,11H2,1-3H3/t15-,16+,17-,18+,19+,20-,21+/m0/s1 |
| SMILES | CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@H](O)C[C@]12C |
| IUPAC Name | (8S,9S,10R,11R,13S,14S,17S)-17-acetyl-11-hydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction