missing translation for 'onlineSavingsMsg'
Learn More

11-Oxo-1,4,7,10-tetraazabicyclo[8.2.2]tetradecane-4,7-diacetic Acid Ammonia salt, TRC

Product Code. 30426211
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
This item is not returnable. View return policy

Product Code. 30426211

Brand: Toronto Research Chemicals TRCO8716205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Oxo-1,4,7,10-tetraazabicyclo[8.2.2]tetradecane-4,7-diacetic Acid Ammonia salt
Analyte or Component Names 11-Oxo-1,4,7,10-tetraazabicyclo[8.2.2]tetradecane-4,7-diacetic Acid Ammonia salt
Shipping Condition Room Temperature
Molecular Formula C14H24N4O5 • xNH3
InChI Formula InChI=1S/C14H24N4O5.H3N/c19-12-9-15-1-2-16(10-13(20)21)3-4-17(11-14(22)23)6-8-18(12)7-5-15;/h1-11H2,(H,20,21)(H,22,23);1H3
Solution Type Neat
SMILES O=C1CN(CCN2CC(O)=O)CCN1CCN(CC2)CC(O)=O.N
IUPAC Name 2,2'-(11-oxo-1,4,7,10-tetraazabicyclo[8.2.2]tetradecane-4,7-diyl)diacetic acid, ammonia salt
Industry Type Pharmaceutical
Molecular Weight (g/mol) 345.4
Quantity 5 mg
Formula Weight 345.2012
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.