missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen 1,4-Sorbitan Ester, TRC

Product Code. 30137056
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137056 10 mg 10mg
30137673 25 mg 25mg
30007213 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137056 Supplier Toronto Research Chemicals Supplier No. TRCK20270010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Analyte or Component Names Ketoprofen 1,4-Sorbitan Ester
Industry Type Forensics

Specifications

Shipping Condition Room Temperature
Molecular Formula C22H24O7
Synonym (R)-2-((2R,3R,4S)-3,4-Dihydroxytetrahydrofuran-2-yl)-2-hydroxyethyl 2-(3-benzoylphenyl)propanoate;
InChI Formula InChI=1S/C22H24O7/c1-13(22(27)29-12-18(24)21-20(26)17(23)11-28-21)15-8-5-9-16(10-15)19(25)14-6-3-2-4-7-14/h2-10,13,17-18,20-21,23-24,26H,11-12H2,1H3/t13?,17-,18+,20+,21+/m0/s1
SMILES O=C(C1=CC=CC=C1)C2=CC(C(C(OC[C@@H](O)[C@@]3([H])OC[C@H](O)[C@H]3O)=O)C)=CC=C2
Molecular Weight (g/mol) 400.42
Quantity 10 mg
Chemical Name or Material Ketoprofen 1,4-Sorbitan Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.