missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-LSD, Lipomed
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Iso-LSD
Specifications
Specifications
| Chemical Name or Material | Iso-LSD (Iso-Lysergic Acid Diethylamide) |
| CAS | 2126-78-5 |
| Analyte or Component Names | Iso-LSD (Iso-Lysergic Acid Diethylamide) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N3 O |
| Synonym | Ergoline-8-carboxamide, 9,10-didehydro-N,N-diethyl-6-methyl-, (8α)-, Isolysergamide, N,N-diethyl- (6 CI,7 CI), (8α)-9,10-Didehydro-N,N-diethyl-6-methylergoline-8-carboxamide, Indolo[4,3-fg]quinoline, ergoline-8-carboxamide deriv., (+)-Isolysergic acid diethylamide, D-Iso-LSD, D-Isolysergic acid diethylamide, D-iso-Lysergic acid N,N-diethylamide, Iso-LSD, Iso-lysergide, Isolysergic acid diethylamide, N,N-Diethylisolysergamide, iso-LSD |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCN(CC)C(=O)[C@@H]1 CN(C)[C@@H]2 Cc3c[nH]c4cccc(C2=C1)c34 |
| IUPAC Name | (6 aR,9 S)-N,N-diethyl-7-methyl-6,6 A,8,9-tetrahydro-4 H-indolo[4,3-fg]quinoline-9-carboxamide |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction