missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Indapamide, TRC

Product Code. 17983955
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17983955 10 mg 10mg
30137228 50 mg 50mg
30136620 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983955 Supplier Toronto Research Chemicals Supplier No. TRCD22966010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63968-75-2
Molecular Formula C16 H14 Cl N3 O3 S
Molecular Weight (g/mol) 363.82
Synonym 4-Chloro-N-(2-methyl-1H-indol-1-yl)-3-sulphamoylbenzamide, Indapamide Imp. B (EP)
IUPAC Name 4-chloro-N-(2-methylindol-1-yl)-3-sulfamoylbenzamide
SMILES Cc1cc2ccccc2n1NC(=O)c3ccc(Cl)c(c3)S(=O)(=O)N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H14ClN3O3S/c1-10-8-11-4-2-3-5-14(11)20(10)19-16(21)12-6-7-13(17)15(9-12)24(18,22)23/h2-9H,1H3,(H,19,21)(H2,18,22,23)
Formula Weight 363.0444
Percent Purity >95%
Chemical Name or Material 4-chloro-3-sulfamoyl-N-(2-methyl-1H-indol-1-yl)benzamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.