CAS RN 63968-75-2
CAS RN 63968-75-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Dehydro Indapamide, TRC
CAS: 63968-75-2 Molecular Formula: C16 H14 Cl N3 O3 S Molecular Weight (g/mol): 363.82 Synonym: 4-Chloro-N-(2-methyl-1H-indol-1-yl)-3-sulphamoylbenzamide,Indapamide Imp. B (EP) IUPAC Name: 4-chloro-N-(2-methylindol-1-yl)-3-sulfamoylbenzamide SMILES: Cc1cc2ccccc2n1NC(=O)c3ccc(Cl)c(c3)S(=O)(=O)N
2
4-Chloro-N-(2-methyl-1H-indol-1-yl)-3-sulphamoylbenzamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.