missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbic Acid (L-Ascorbic Acid; Vitamin C), LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Ascorbic Acid (L-Ascorbic Acid; Vitamin C)
Specifications
Specifications
| Chemical Name or Material | L-Ascorbic Acid |
| CAS | 50-81-7 |
| Analyte or Component Names | L-Ascorbic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H8 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Ascorbic Acid, (+)-Ascorbic acid, 100 M, 3-Oxo-L-gulofuranolactone, 3-keto-L-Gulofuranolactone, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M, Viscorin 100 M |
| InChI Formula | InChI=1 S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10 H,1H2/t2-,5+/m0/s1 |
| SMILES | OC[C@H](O)[C@H]1 OC(=O)C(=C1O)O |
| IUPAC Name | (2 R)-2-[(1 S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2 H-furan-5-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction