missing translation for 'onlineSavingsMsg'
Learn More

TLE3 Antibody (CL3573) - Azide and BSA Free, Novus Biologicals™

Código de producto. 30296381 Tienda Bio Techne Productos
Change view
Click to view available options
Quantity:
100 μL
Tamaño de la unidad:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
30296381 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30296381 Proveedor Novus Biologicals N.º de proveedor NBP343936

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

TLE3 Monoclonal antibody specifically detects TLE3 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence,Immunohistochemistry,KnockDown
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen TLE3
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, KnockDown
Classification Monoclonal
Clone CL3573
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:10000 - 1:20000
Formulation Lyophilized in 5% Trehalose.
Gene Alias ESG3Enhancer of split groucho-like protein 3, ESGenhancer of split groucho 3, GRG3, HsT18976, KIAA1547FLJ39460, transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 3, homolog of Drosophila E(sp1), transducin-like enhancer protein 3
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q04726, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7090
Reconstitution Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Target Species Human
Content And Storage Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.