missing translation for 'onlineSavingsMsg'
Få mere at vide

Novus Biologicals™ TRAF-1 Antibody (TRAF1/3365), Novus Biologicals™

Artikelnummer. 30043944 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
100 μg
20 μg
Pakningsstørrelse:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30043944 100 μg 100µg
30043443 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30043944 Leverandør Novus Biologicals™ Leverandørnr. NBP326741100ug

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

TRAF-1 Monoclonal antibody specifically detects TRAF-1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen TRAF-1
Applications ELISA
Classification Monoclonal
Clone TRAF1/3365
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 10 mM PBS with 0.05% BSA
Gene Alias EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1
Host Species Mouse
Immunogen Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 7185
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.