missing translation for 'onlineSavingsMsg'
Learn More

TRIP6 Antibody (2G2), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43054

Product Code. 30279501

  • 408.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRIP6 Monoclonal antibody specifically detects TRIP6 in Human,Mouse samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIP6
Monoclonal
Unconjugated
PBS, pH 7.3, 50% glycerol
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Western Blot
2G2
Western Blot 1:500 - 1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:20 - 1:200
MGC10556, MGC10558, MGC29959, MGC3837, MGC4423, OIP1OIP-1, Opa-interacting protein 1, thyroid hormone receptor interactor 6, thyroid receptor-interacting protein 6, TR-interacting protein 6, ZRP-1TRIP-6, zyxin related protein 1, Zyxin-related protein 1
thyroid hormone receptor interactor 6 - (Uniprot# Q15654)
100 μg
Core ESC Like Genes, Stem Cell Markers
7205
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.