Learn More
NDUFS5 Antibody (5C2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43006
This item is not returnable.
View return policy
Description
NDUFS5 Monoclonal antibody specifically detects NDUFS5 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| NDUFS5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 5C2 | |
| Western Blot 1:1000 - 1:4000, ELISA, Immunohistochemistry 1:20 - 1200 | |
| CI-15 kDa, CI-15k, CI15K, Complex I-15 kDa, NADH dehydrogenase (ubiquinone) Fe-S protein 5 (15kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 5, 15kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 5, NADH:ubiquinone oxidoreductase 15 kDa IP subunit, NADH-ubiquinone oxidoreductase 15 kDa subunit | |
| NADH dehydrogenase(ubiquinone) Fe-S protein 5, 15kDa(NADH-coenzyme Q reductase) - (Uniprot# O43920) | |
| 100 μg | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4725 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |